2,4-Dimethyl-3-pentanone 98%

Supplier: Thermo Scientific Chemicals
Danger

A12875-22 A12875-36
CAAAA12875-22EA 65.41 CAD
CAAAA12875-22 CAAAA12875-36
2,4-Dimethyl-3-pentanone 98%
2,4-Dimethyl-3-pentanone

Purity: 98% CAS No.: 565-80-0 Molecular Formula: C7H14O Formula Weight: 114.19 Flash Point: 15°(59°F) Refractive Index: 1.3990 Melting Point: -69° Boiling Point: 124-125° Beilstein Reference: 773782 UN No.: 1224 MDL No.: MFCD00008918

Formula: C₇H₁₄O
Boiling Pt: 124…125 °C
Melting Pt: –69 °C
Density: 0.803
Flash Pt: 15 °C (59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008918
CAS Number: 565-80-0
EINECS: 209-294-7
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR