4-Chloro-2-nitrobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

A12872-14 A12872-22
CAAAA12872-14EA 24.02 CAD
CAAAA12872-14 CAAAA12872-22
4-Chloro-2-nitrobenzoic acid 97%
4-Chloro-2-nitrobenzoic acid

Purity: 97% CAS No.: 6280-88-2 Molecular Formula: C7H4ClNO4 Formula Weight: 201.57 Flash Point: 100°(212°F) Odor: Odorless Beilstein Reference: 2051370 MDL No.: MFCD00007214

Formula: C₇H₄ClNO₄
Melting Pt: 142…146 °C
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007214
CAS Number: 6280-88-2
EINECS: 228-483-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR