4-Bromobutyric acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Bromobutanoic acid
A12870-09
A12870-18
A12870-30
CAAAA12870-09EA
67.94
CAD
CAAAA12870-09
CAAAA12870-18
CAAAA12870-30
4-Bromobutyric acid 97%
4-Bromobutyric acid
Purity: 97%
CAS No.: 2623-87-2
Molecular Formula: Br(CH2)3CO2H
Formula Weight: 167.01
Flash Point: 115°(239°F)
Odor: Stench
Boiling Point: 129-131°/11mm
Beilstein Reference: 1743056
UN No.: 3261
MDL No.: MFCD00002817
Formula:
BrH₂C(CH₂)₂COOH MW: 167 g/mol Boiling Pt: 129…131 °C/11 mm Melting Pt: 31…33 °C Flash Pt: 115 °C (239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002817 CAS Number: 2623-87-2 EINECS: 220-083-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...