2,4-Dihydroxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

A12868-36 A12868-14 A12868-22
CAAAAA12868-36EA 1309.82 CAD
CAAAAA12868-36 CAAAA12868-14 CAAAA12868-22
2,4-Dihydroxybenzaldehyde 98%
2,4-Dihydroxybenzaldehyde

Purity: 98% CAS No.: 95-01-2 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 134-138° Boiling Point: 220°/22mm Merck Reference: 14,8156 MDL No.: MFCD00011686

Formula: C₇H₆O₃
Boiling Pt: 220 °C/22 mm
Melting Pt: 134…138 °C
Density: 1.448 g/cm³ (-93 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011686
CAS Number: 95-01-2
EINECS: 202-383-1
Merck Index: 14,8156

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR