Trimethylsilylacetylene 98%

Supplier: Thermo Scientific Chemicals
Danger

A12856-06 A12856-14 A12856-22
CAAAA12856-06EA 83.27 CAD
CAAAA12856-06 CAAAA12856-14 CAAAA12856-22
Trimethylsilylacetylene 98%
Trimethylsilylacetylene

Purity: 98% CAS No.: 1066-54-2 Molecular Formula: C5H10Si Formula Weight: 98.22 Flash Point: -34°(-29°F) Refractive Index: 1.3900 Boiling Point: 52-53° Beilstein Reference: 906752 UN No.: 1993 MDL No.: MFCD00008569

Formula: C₅H₁₀Si
Boiling Pt: 52…53 °C
Density: 0.710
Flash Pt: <–34 °C (–29 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008569
CAS Number: 1066-54-2
EINECS: 213-919-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR