2,6-Dichlorothiophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12854-06
A12854-14
CAAAAA12854-22EA
1138.71
CAD
CAAAAA12854-22
CAAAA12854-06
CAAAA12854-14
2,6-Dichlorothiophenol 97%
2,6-Dichlorothiophenol
Purity: 97%
CAS No.: 24966-39-0
Molecular Formula: C6H4Cl2S
Formula Weight: 179.07
Flash Point: 112°(233°F)
Odor: Unpleasant
Boiling Point: 101-103°/14mm
Beilstein Reference: 2082400
MDL No.: MFCD00004833
Formula:
C₆H₄Cl₂S Boiling Pt: 101…103 °C (14 mmHg) Melting Pt: 46…50 °C Flash Pt: 112 °C (233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004833 CAS Number: 24966-39-0 EINECS: 416-720-7 UN: 3335 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...