2-Hydroxychalcone 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(2-Hydroxyphenyl)-1-phenylprop-2-en-1-one, 2-Hydroxybenzylidenacetophenone

A12851-06 A12851-14
CAAAA12851-06EA 62.88 CAD
CAAAA12851-06 CAAAA12851-14
2-Hydroxychalcone 98+%
2-Hydroxychalcone

Purity: 98+% CAS No.: 644-78-0 Molecular Formula: C15H12O2 Formula Weight: 224.26 Melting Point: ca 154° dec. Beilstein Reference: 2049134

Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
Melting Pt: ∼ 154 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00016449
CAS Number: 644-78-0
EINECS: 211-422-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR