Potassium sorbate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate

A12844-0B A12844-0I A12844-30
CAAAA12844-0BEA 247.29 CAD
CAAAA12844-0B CAAAA12844-0I CAAAA12844-30
Potassium sorbate 99%
Potassium sorbate

Purity: 99% CAS No.: 24634-61-5 Molecular Formula: C6H7KO2 Formula Weight: 150.22 pH: 8,3 Melting Point: 300° MDL No.: MFCD00016546

Formula: CH₃CH=CHCH=CHCOOK
MW: 150.22 g/mol
Melting Pt: >300 °C
Density: 1.361
Storage Temperature: Ambient
MDL Number: MFCD00016546
CAS Number: 24634-61-5
EINECS: 246-376-1
Merck Index: 14,7671
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500421971592bd8f37c1a6b88f97c1de033cbf87&vid=e8c4a1d0da78946f174af6a94e5c78fc9c16de05&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR