5-Chlorovanillic acid tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12841-06
A12841-14
CAAAA12841-06EA
101.44
CAD
CAAAA12841-06
CAAAA12841-14
CAAAA12841-22
5-Chlorovanillic acid tech. 90%, Technical Grade
5-Chlorovanillic acid
Purity: 90%
CAS No.: 62936-23-6
Molecular Formula: C8H7ClO4
Formula Weight: 202.60
Melting Point: 242-248°
Beilstein Reference: 2648897
Formula:
C₈H₇ClO₄ Melting Pt: 242…248 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016531 CAS Number: 62936-23-6 EINECS: 263-766-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...