2-Pyrrolecarbaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12838-14
A12838-22
A12838-09
CAAAAA12838-14EA
270.18
CAD
CAAAAA12838-14
CAAAAA12838-22
CAAAA12838-09
2-Pyrrolecarbaldehyde 99%
2-Pyrrolecarbaldehyde
Purity: 99%
CAS No.: 1003-29-8
Molecular Formula: C5H5NO
Formula Weight: 95.10
Storage Temperature: 2°C to 8°C
Flash Point: 106°(222°F)
Melting Point: 40-46°
Boiling Point: 217-219°
Beilstein Reference: 105745
MDL No.: MFCD00005217
Formula:
C₅H₅NO Boiling Pt: 217…219 °C Melting Pt: 40…46 °C Flash Pt: 106 °C (222 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005217 CAS Number: 1003-29-8 EINECS: 213-705-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...