2,5-Dimethylfuran 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12833-14
A12833-22
A12833-36
CAAAA12833-14EA
75.05
CAD
CAAAA12833-14
CAAAA12833-22
CAAAA12833-36
2,5-Dimethylfuran 98+%
2,5-Dimethylfuran
Purity: 98+%
CAS No.: 625-86-5
Molecular Formula: C6H8O
Formula Weight: 96.13
Flash Point: 1°(33°F)
Refractive Index: 1.4410
Melting Point: -62°
Boiling Point: 92-93°
Beilstein Reference: 106449
UN No.: 3271
MDL No.: MFCD00003250
Formula:
C₆H₈O Boiling Pt: 92…93 °C Melting Pt: –62 °C Density: 0.903 Flash Pt: 1 °C (33 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003250 CAS Number: 625-86-5 EINECS: 210-914-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...