9,10-Dichloroanthracene 97%

Supplier: Thermo Scientific Chemicals
Warning

A12823-06 A12823-14
CAAAA12823-06EA 92.9 CAD
CAAAA12823-06 CAAAA12823-14
9,10-Dichloroanthracene 97%
9,10-Dichloroanthracene

Purity: 97% CAS No.: 605-48-1 Molecular Formula: C14H8Cl2 Formula Weight: 247.12 Melting Point: 210-215° Beilstein Reference: 1912108

Formula: C₁₄H₈Cl₂
Boiling Pt: 366 °C (1013 hPa)
Melting Pt: 210…215 °C
Density: 1,525 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001246
CAS Number: 605-48-1
EINECS: 210-087-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR