1,4-Dimethoxybenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12822-0E
A12822-22
A12822-36
CAAAA12822-0EEA
443.98
CAD
CAAAA12822-0E
CAAAA12822-22
CAAAA12822-36
1,4-Dimethoxybenzene 98%
1,4-Dimethoxybenzene
Purity: 98%
CAS No.: 150-78-7
Molecular Formula: C8H10O2
Formula Weight: 138.17
Flash Point: 94°(201°F)
Odor: Sweet
Boiling Point: 212°
Beilstein Reference: 774605
MDL No.: MFCD00008401
Formula:
C₈H₁₀O₂ Boiling Pt: 212 °C Melting Pt: 54…58 °C Density: 1,053 Flash Pt: 94 °C(201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008401 CAS Number: 150-78-7 EINECS: 205-771-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...