n-Hexylbenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12821-14
A12821-22
A12821-36
CAAAA12821-14EA
121.03
CAD
CAAAA12821-14
CAAAA12821-22
CAAAA12821-36
n-Hexylbenzene 98%
n-Hexylbenzene
Purity: 98%
CAS No.: 1077-16-3
Molecular Formula: C12H18
Formula Weight: 162.28
Flash Point: 83°(181°F)
Refractive Index: 1.4860
Melting Point: -61°
Boiling Point: 226-227°
Beilstein Reference: 1904786
UN No.: 3082
MDL No.: MFCD00009526
Formula:
C₁₂H₁₈ Boiling Pt: 226…227 °C Melting Pt: –61 °C Density: 0.861 Flash Pt: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009526 CAS Number: 1077-16-3 EINECS: 214-070-7 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...