2-Chloro-1-methylpyridinium iodide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12820-36
A12820-14
A12820-22
CAAAAA12820-36EA
1245.04
CAD
CAAAAA12820-36
CAAAA12820-14
CAAAA12820-22
2-Chloro-1-methylpyridinium iodide 97%
2-Chloro-1-methylpyridinium iodide
Purity: 97%
CAS No.: 14338-32-0
Molecular Formula: C6H7ClIN
Formula Weight: 255.49
Melting Point: ca 200° dec.
Beilstein Reference: 3572320
Merck Reference: 14,6301
MDL No.: MFCD00011984
Formula:
C₆H₇ClIN Melting Pt: ∼ 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011984 CAS Number: 14338-32-0 EINECS: 238-288-7 Merck Index: 14,6301 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...