Bromocyclobutane 95%

Supplier: Thermo Scientific Chemicals
Danger

A12817-14 A12817-03 A12817-06
CAAAAA12817-14EA 1756.96 CAD
CAAAAA12817-14 CAAAA12817-03 CAAAA12817-06
Bromocyclobutane 95%
Bromocyclobutane

Purity: 95% CAS No.: 4399-47-7 Molecular Formula: C4H7Br Formula Weight: 135.01 Flash Point: 22°(71°F) Refractive Index: 1.4795 Boiling Point: 107-108° Beilstein Reference: 1900415 UN No.: 1993 MDL No.: MFCD00001317

Formula: C₄H₇Br
Boiling Pt: 107…108 °C
Density: 1.431
Flash Pt: 22 °C (71 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001317
CAS Number: 4399-47-7
EINECS: 224-530-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR