4-Butylphenol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12812-06
A12812-14
A12812-22
CAAAA12812-06EA
113.44
CAD
CAAAA12812-06
CAAAA12812-14
CAAAA12812-22
4-Butylphenol 98%
4-Butylphenol
Purity: 98%
CAS No.: 1638-22-8
Molecular Formula: C10H14O
Formula Weight: 150.22
Refractive Index: 1.5170
Melting Point: ca 22°
Boiling Point: 245°
Beilstein Reference: 2042120
UN No.: 3145
MDL No.: MFCD00041750
Formula:
C₁₀H₁₄O Boiling Pt: 245 °C Melting Pt: ∼ 22 °C Density: 0.980 Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041750 CAS Number: 1638-22-8 EINECS: 216-672-5 UN: 3145 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...