2,6-Dimethyl-4-pyrone 99%

Supplier: Thermo Scientific Chemicals
Warning

A12799-09 A12799-18 A12799-30
CAAAA12799-09EA 80.11 CAD
CAAAA12799-09 CAAAA12799-18 CAAAA12799-30
2,6-Dimethyl-4-pyrone 99%
2,6-Dimethyl-4-pyrone

Purity: 99% CAS No.: 1004-36-0 Molecular Formula: C7H8O2 Formula Weight: 124.14 Melting Point: 132-136° Boiling Point: 248-250° MDL No.: MFCD00006579

Formula: C₇H₈O₂
Boiling Pt: 248…250 °C
Melting Pt: 132…136 °C
Density: 1.17 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006579
CAS Number: 1004-36-0
EINECS: 213-719-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR