5-Fluoroindole-2-carboxylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A12794-03 A12794-06 A12794-14
CAAAA12794-03EA 65.73 CAD
CAAAA12794-03 CAAAA12794-06 CAAAA12794-14
5-Fluoroindole-2-carboxylic acid 98+%
5-Fluoroindole-2-carboxylic acid

Purity: 98+% CAS No.: 399-76-8 Molecular Formula: C9H6FNO2 Formula Weight: 179.15 Melting Point: ca 259° dec. Beilstein Reference: 153217

Formula: C₉H₆FNO₂
Melting Pt: ∼ 259 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005612
CAS Number: 399-76-8
EINECS: 206-919-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR