2-Bromo-4-fluorotoluene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12792-22
A12792-06
A12792-14
CAAAAA12792-22EA
796.32
CAD
CAAAAA12792-22
CAAAA12792-06
CAAAA12792-14
2-Bromo-4-fluorotoluene 98+%
2-Bromo-4-fluorotoluene
Purity: 98+%
CAS No.: 1422-53-3
Molecular Formula: C7H6BrF
Formula Weight: 189.03
Flash Point: 64°(147°F)
Refractive Index: 1.5280
Boiling Point: 170-172°
Beilstein Reference: 1859038
MDL No.: MFCD00040828
Formula:
C₇H₆BrF Boiling Pt: 170…172 °C Density: 1,526 Flash Pt: 64 °C(147 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040828 CAS Number: 1422-53-3 EINECS: 215-830-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...