4'-(Trifluoromethyl)acetanilide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12789-06
A12789-14
CAAAAA12789-22EA
1438.83
CAD
CAAAAA12789-22
CAAAA12789-06
CAAAA12789-14
4'-(Trifluoromethyl)acetanilide 98+%
4'-(Trifluoromethyl)acetanilide
Purity: 98+%
CAS No.: 349-97-3
Molecular Formula: C9H8F3NO
Formula Weight: 203.16
Odor: Odorless
Melting Point: 150-154°
Beilstein Reference: 2106965
MDL No.: MFCD00013562
Formula:
C₉H₈F₃NO Melting Pt: 150…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013562 CAS Number: 349-97-3 EINECS: 206-495-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...