Iodobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Phenyliodide
A12788-22
A12788-0B
A12788-18
CAAAAA12788-22EA
116.29
CAD
CAAAAA12788-22
CAAAA12788-0B
CAAAA12788-18
Iodobenzene 98%
Iodobenzene
Purity: 98%
CAS No.: 591-50-4
Molecular Formula: C6H5I
Formula Weight: 204.01
Flash Point: 74°(165°F)
Physical Form: Liquid
Density: 1.826
Refractive Index: 1.6200
Sensitivity: Light Sensitive
Appearance: Colorless to pale yellow
Odor: Aromatic
Melting Point: -31°
Boiling Point: 188-189°
Beilstein Reference: 1446140
Merck Reference: 14,5029
MDL No.: MFCD00001029
Formula:
C₆H₅I MW: 204.01 g/mol Boiling Pt: 188…189 °C Melting Pt: –31 °C Density: 1.826 Flash Pt: 74 °C(165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001029 CAS Number: 591-50-4 EINECS: 209-719-6 Merck Index: 14,5029 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...