Difluoroacetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12787-06
A12787-14
CAAAA12787-06EA
131.46
CAD
CAAAA12787-06
CAAAA12787-14
Difluoroacetic acid 98%
Difluoroacetic acid
Purity: 98%
CAS No.: 381-73-7
Molecular Formula: F2CHCO2H
Formula Weight: 96.03
Flash Point: 78°(172°F)
Refractive Index: 1.3430
Odor: Pungent
Melting Point: -1°
Boiling Point: 132-134°
Beilstein Reference: 1098588
UN No.: 3265
MDL No.: MFCD00004220
Formula:
C₂H₂F₂O₂ Boiling Pt: 132…134 °C Melting Pt: –1 °C Density: 1.526 Flash Pt: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004220 CAS Number: 381-73-7 EINECS: 206-839-0 UN: 3265 ADR: 8,III |
Specification Test Results
Refractive index | 1.3430 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...