α,α,α-Trifluoro-m-cresol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12784-06
A12784-14
A12784-22
CAAAA12784-06EA
37.29
CAD
CAAAA12784-06
CAAAA12784-14
CAAAA12784-22
α,α,α-Trifluoro-m-cresol 98+%
α,α,α-Trifluoro-m-cresol
Purity: 98+%
CAS No.: 98-17-9
Molecular Formula: C7H5F3O
Formula Weight: 162.11
Flash Point: 74°(165°F)
Refractive Index: 1.4580
Odor: Phenol-like
Melting Point: -2°
Boiling Point: 177-178°
Beilstein Reference: 2045663
UN No.: 3265
MDL No.: MFCD00002299
Formula:
C₇H₅F₃O Boiling Pt: 177…178 °C Melting Pt: –2 °C Density: 1.336 Flash Pt: 74 °C (165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002299 CAS Number: 98-17-9 EINECS: 202-645-5 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...