2-Fluoromesitylene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluoro-1,3,5-trimethylbenzene, Fluoromesitylene, 2,4,6-Trimethylfluorobenzene
A12779-03
A12779-06
CAAAA12779-03EA
88.48
CAD
CAAAA12779-03
CAAAA12779-06
2-Fluoromesitylene 98%
2-Fluoromesitylene
Purity: 98%
CAS No.: 392-69-8
Molecular Formula: C9H11F
Formula Weight: 138.19
Refractive Index: 1.4830
Boiling Point: 163-165°
MDL No.: MFCD00052365
Formula:
C₉H₁₁F MW: 138.18 g/mol Boiling Pt: 163…165 °C Density: 0.975 Storage Temperature: Ambient |
MDL Number:
MFCD00052365 CAS Number: 392-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...