Metacetamol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12764-14
A12764-22
CAAAAA12764-14EA
49.93
CAD
CAAAAA12764-14
CAAAA12764-22
CAAAA12764-36
Metacetamol 99%
Metacetamol
Purity: 99%
CAS No.: 621-42-1
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 146-149°
Beilstein Reference: 907998
Formula:
C₈H₉NO₂ Boiling Pt: 351 °C (1013 hPa) Melting Pt: 146…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002263 CAS Number: 621-42-1 EINECS: 210-687-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...