2-(Methylthio)aniline 98%

Supplier: Thermo Scientific Chemicals
Danger

A12762-06 A12762-14 A12762-22
CAAAA12762-06EA 52.93 CAD
CAAAA12762-06 CAAAA12762-14 CAAAA12762-22
2-(Methylthio)aniline 98%
2-(Methylthio)aniline

Purity: 98% CAS No.: 2987-53-3 Molecular Formula: C7H9NS Formula Weight: 139.23 Flash Point: 110°(230°F) Refractive Index: 1.6240 Boiling Point: 233-234° Beilstein Reference: 386211 UN No.: 2810 MDL No.: MFCD00007708

Formula: C₇H₉NS
Boiling Pt: 233…234 °C
Density: 1.111
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007708
CAS Number: 2987-53-3
EINECS: 221-055-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR