4-Chlorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

A12757-0B A12757-0I A12757-30
CAAAA12757-0BEA 240.16 CAD
CAAAA12757-0B CAAAA12757-0I CAAAA12757-30
4-Chlorobenzaldehyde 98%
4-Chlorobenzaldehyde

Purity: 98% CAS No.: 104-88-1 Molecular Formula: C7H5ClO Formula Weight: 140.57 Flash Point: 88°(190°F) Odor: Pungent Boiling Point: 213-214° Beilstein Reference: 385858 UN No.: 3077 MDL No.: MFCD00003379

Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 213…214 °C
Melting Pt: 45…49 °C
Density: 1.196 g/cm³ (20 °C)
Flash Pt: 88 °C(190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003379
CAS Number: 104-88-1
EINECS: 203-247-4
UN: 3077
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR