Benzocaine (ethyl-4-aminobenzoate) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-aminobenzoate, Aethoform

A12754-0I A12754-0B A12754-30
CAAAAA12754-0IEA 1117.06 CAD
CAAAAA12754-0I CAAAA12754-0B CAAAA12754-30
Benzocaine (ethyl-4-aminobenzoate) 98%
Benzocaine (Ethyl 4-aminobenzoate)

Purity: 98% CAS No.: 94-09-7 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Flash Point: 110°(230°F) Odor: Odorless Boiling Point: 310° Beilstein Reference: 638434 Merck Reference: 14,1086 MDL No.: MFCD00007892

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 310 °C
Melting Pt: 88…92 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007892
CAS Number: 94-09-7
EINECS: 202-303-5
Merck Index: 14,1086

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR