1-Dodecanethiol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Mercaptan C12, 1-Mercaptododecane, n-Dodecanethiol, Lauryl mercaptan, n-Dodecyl mercaptan, NDM
A12741-22
A12741-36
CAAAA12741-22EA
46.14
CAD
CAAAA12741-22
CAAAA12741-36
1-Dodecanethiol 98%
1-Dodecanethiol
Purity: 98%
CAS No.: 112-55-0
Molecular Formula: C12H26S
Formula Weight: 202.40
Flash Point: 133°(271°F)
Refractive Index: 1.4601
Odor: Unpleasant
Melting Point: -7°
Boiling Point: 276-278°
Beilstein Reference: 969337
MDL No.: MFCD00004885
Formula:
C₁₂H₂₆S MW: 202.4 g/mol Boiling Pt: 276…278 °C Melting Pt: –7 °C Density: 0.845 Flash Pt: 133 °C (271 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004885 CAS Number: 112-55-0 EINECS: 203-984-1 UN: 1760 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...