Oxalacetic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Oxaloacetic acid, 2-Succinic acid, Oxalacetic acid

A12739-06 A12739-14 A12739-22
CAAAA12739-06EA 161.16 CAD
CAAAA12739-06 CAAAA12739-14 CAAAA12739-22
Oxalacetic acid 97%
Oxalacetic acid

Purity: 97% CAS No.: 328-42-7 Molecular Formula: C4H4O5 Formula Weight: 132.07 Storage Temperature: -30°C to -10°C Flash Point: 88°(190°F) Melting Point: 161° dec. Beilstein Reference: 1705475 Merck Reference: 14,6909 UN No.: 3261 MDL No.: MFCD00002592

Formula: C₄H₄O₅
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 161 °C (dec.)
Flash Pt: 88 °C (190 °F)
Storage Temperature: Freezer
MDL Number: MFCD00002592
CAS Number: 328-42-7
EINECS: 206-329-8
UN: 3261
ADR: 8,III
Merck Index: 14,6909

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR