(3-Fluorophenyl)acetonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

A12737-06 A12737-14
CAAAA12737-06EA 57.83 CAD
CAAAA12737-06 CAAAA12737-14
(3-Fluorophenyl)acetonitrile 99%
(3-Fluorophenyl)acetonitrile

Purity: 99% CAS No.: 501-00-8 Molecular Formula: C8H6FN Formula Weight: 135.14 Flash Point: 110°(230°F) Refractive Index: 1.5020 Boiling Point: 230° Beilstein Reference: 1861071 UN No.: 3276 MDL No.: MFCD00001907

Formula: C₈H₆FN
Boiling Pt: 230 °C
Density: 1.156
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001907
CAS Number: 501-00-8
EINECS: 207-918-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR