α,3,5-Tribromotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12726-14
A12726-03
A12726-06
CAAAAA12726-14EA
1440.96
CAD
CAAAAA12726-14
CAAAA12726-03
CAAAA12726-06
α,3,5-Tribromotoluene 99%
α,3,5-Tribromotoluene
Purity: 99%
CAS No.: 56908-88-4
Molecular Formula: C7H5Br3
Formula Weight: 328.84
Melting Point: 93-95°
Beilstein Reference: 3237827
UN No.: 1759
MDL No.: MFCD00052415
Formula:
C₇H₅Br₃ Boiling Pt: 173 °C (19 -rr) Melting Pt: 93…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052415 CAS Number: 56908-88-4 UN: 2921 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...