4-(Trifluoromethyl)thiophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Mercaptobenzotrifluoride, 4-(Trifluoromethyl)benzene-1-thiol, 4-(Trifluoromethyl)benzenethiol
A12725-03
A12725-06
CAAAAA12725-14EA
4772.33
CAD
CAAAAA12725-14
CAAAA12725-03
CAAAA12725-06
4-(Trifluoromethyl)thiophenol 97%
4-(Trifluoromethyl)thiophenol
Purity: 97%
CAS No.: 825-83-2
Molecular Formula: C7H5F3S
Formula Weight: 178.18
Refractive Index: 1.4920
Odor: Stench
Boiling Point: 66-68°/15mm
Beilstein Reference: 2045665
UN No.: 2810
MDL No.: MFCD00042320
Formula:
C₇H₅F₃S MW: 178.18 g/mol Boiling Pt: 66…68 °C (15 mmHg) Density: 1.300 Storage Temperature: Ambient |
MDL Number:
MFCD00042320 CAS Number: 825-83-2 UN: 2810 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...