1,3-Dibromobenzene 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12723-14
A12723-22
A12723-36
CAAAA12723-14EA
111.39
CAD
CAAAA12723-14
CAAAA12723-22
CAAAA12723-36
1,3-Dibromobenzene 97+%
1,3-Dibromobenzene
Purity: 97+%
CAS No.: 108-36-1
Molecular Formula: C6H4Br2
Formula Weight: 235.91
Flash Point: 93°(199°F)
Refractive Index: 1.6080
Odor: Odorless
Melting Point: -7°
Boiling Point: 219-220°
Beilstein Reference: 1904538
UN No.: 3082
MDL No.: MFCD00000078
Formula:
C₆H₄Br₂ Boiling Pt: 219…220 °C Melting Pt: –7 °C Density: 1.955 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000078 CAS Number: 108-36-1 EINECS: 203-574-2 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...