2-Amino-3,5-dibromopyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12706-22
A12706-14
CAAAAA12706-22EA
524.56
CAD
CAAAAA12706-22
CAAAA12706-06
CAAAA12706-14
2-Amino-3,5-dibromopyridine 97%
2-Amino-3,5-dibromopyridine
Purity: 97%
CAS No.: 35486-42-1
Molecular Formula: C5H4Br2N2
Formula Weight: 251.92
Melting Point: 102-105°
Beilstein Reference: 119390
Formula:
C₅H₄Br₂N₂ Boiling Pt: 254 °C (1013 hPa) Melting Pt: 102…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038041 CAS Number: 35486-42-1 EINECS: 252-590-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...