2-Chloro-6-nitrobenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12703-06
A12703-14
CAAAA12703-06EA
51.98
CAD
CAAAA12703-06
CAAAA12703-14
2-Chloro-6-nitrobenzonitrile 98%
2-Chloro-6-nitrobenzonitrile
Purity: 98%
CAS No.: 6575-07-1
Molecular Formula: C7H3ClN2O2
Formula Weight: 182.57
Melting Point: 118-124°
Beilstein Reference: 1369781
Formula:
C₇H₃ClN₂O₂ Melting Pt: 118…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007203 CAS Number: 6575-07-1 EINECS: 229-496-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...