Amantadine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Adamantylamine hydrochloride, 1-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 1-Aminotricyclo(3.3.1.1(3,7))decan HCl, 1-Aminoadamantane hydrochloride, 1-Adamantanammonium chloride, 1-Adamantanamine hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-1-ylamine hydrochloride, Adamantan-1-amine hydrochloride, 1-Adamantaneammonium Chloride

A12699-14 A12699-22 A12699-36
CAAAA12699-14EA 109.49 CAD
CAAAA12699-14 CAAAA12699-22 CAAAA12699-36
Amantadine hydrochloride 99%
Amantadine hydrochloride

Antiviral drug that inhibits ion channels of influenza and disrupts T-cell development


Purity: 99% CAS No.: 665-66-7 Molecular Formula: C10H17N.HCl Formula Weight: 187.71 Melting Point: 360° Beilstein Reference: 4198854 Merck Reference: 14,374 MDL No.: MFCD00074723

Formula: C₁₀H₁₈ClN
MW: 187.71 g/mol
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00074723
CAS Number: 665-66-7
EINECS: 211-560-2
Merck Index: 14,374

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR