2-Methylbenzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12696-09
A12696-18
A12696-30
CAAAA12696-09EA
58.14
CAD
CAAAA12696-09
CAAAA12696-18
CAAAA12696-30
2-Methylbenzyl alcohol 98%
2-Methylbenzyl alcohol
Purity: 98%
CAS No.: 89-95-2
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 104°(219°F)
Refractive Index: 1.5408
Melting Point: 33-37°
Boiling Point: 219°
Beilstein Reference: 1929783
MDL No.: MFCD00004622
Formula:
C₈H₁₀O Boiling Pt: 219 °C Melting Pt: 33…37 °C Density: 1.08 g/cm³ (20 °C) Flash Pt: 104 °C(219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004622 CAS Number: 89-95-2 EINECS: 201-954-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...