4-Chlorophenyl isothiocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12693-06
A12693-14
A12693-22
CAAAA12693-06EA
69.99
CAD
CAAAA12693-06
CAAAA12693-14
CAAAA12693-22
4-Chlorophenyl isothiocyanate 98%
4-Chlorophenyl isothiocyanate
Purity: 98%
CAS No.: 2131-55-7
Molecular Formula: C7H4ClNS
Formula Weight: 169.64
Flash Point:
Formula:
C₇H₄ClNS Boiling Pt: 135…136 °C/24 mm Melting Pt: 43…46 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004810 CAS Number: 2131-55-7 EINECS: 218-358-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...