Hippuric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A12690-22 A12690-36 A12690-0B
CAAAAA12690-22EA 61.94 CAD
CAAAAA12690-22 CAAAAA12690-36 CAAAA12690-0B
Hippuric acid 98%
Hippuric acid

Purity: 98% CAS No.: 495-69-2 Molecular Formula: C9H9NO3 Formula Weight: 179.18 Melting Point: 188-191° Beilstein Reference: 1073987 Merck Reference: 14,4716 MDL No.: MFCD00002692

Formula: C₆H₅C(O)NHCH₂COOH
Boiling Pt: 399 °C (1013 hPa)
Melting Pt: 188…191 °C
Density: 1.371
Storage Temperature: Ambient
MDL Number: MFCD00002692
CAS Number: 495-69-2
EINECS: 207-806-3
Merck Index: 14,4716

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR