trans-2-Pentenoic acid 90+% remainder other isomers
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12689-09
A12689-18
CAAAA12689-09EA
110.76
CAD
CAAAA12689-09
CAAAA12689-18
CAAAA12689-30
trans-2-Pentenoic acid 90+% remainder other isomers
trans-2-Pentenoic acid
Purity: 90+%
CAS No.: 13991-37-2
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 102°(215°F)
Refractive Index: 1.4515
Odor: Pungent
Melting Point: 8-10°
Boiling Point: 200-203°
Beilstein Reference: 1720312
UN No.: 3265
MDL No.: MFCD00002704
Formula:
C₅H₈O₂ Boiling Pt: 200…203 °C Melting Pt: 8…10 °C Density: 0.992 Flash Pt: 102 °C (215 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002704 CAS Number: 13991-37-2 EINECS: 237-791-9 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...