2-Acetylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12688-14
A12688-22
A12688-36
CAAAA12688-14EA
73.79
CAD
CAAAA12688-14
CAAAA12688-22
CAAAA12688-36
2-Acetylpyridine 98%
2-Acetylpyridine
Purity: 98%
CAS No.: 1122-62-9
Molecular Formula: C7H7NO
Formula Weight: 121.14
Flash Point: 76°(169°F)
Refractive Index: 1.5220
Odor: Unpleasant
Melting Point: 8-10°
Boiling Point: 189-190°
Beilstein Reference: 107759
MDL No.: MFCD00006303
Formula:
C₇H₇NO Boiling Pt: 189…190 °C Melting Pt: 8…10 °C Density: 1.082 Flash Pt: 76 °C(169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006303 CAS Number: 1122-62-9 EINECS: 214-355-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...