2,3,5,6-Tetrafluorophenol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12686-06
A12686-14
CAAAA12686-06EA
79.16
CAD
CAAAA12686-06
CAAAA12686-14
2,3,5,6-Tetrafluorophenol 97%
2,3,5,6-Tetrafluorophenol
Purity: 97%
CAS No.: 769-39-1
Molecular Formula: C6H2F4O
Formula Weight: 166.07
Flash Point: 79°(174°F)
Odor: Phenol-like
Boiling Point: 141-143°
Beilstein Reference: 1911548
UN No.: 3261
MDL No.: MFCD00002157
Formula:
C₆H₂F₄O MW: 166.07 g/mol Boiling Pt: 141…143 °C Melting Pt: 28…30 °C Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002157 CAS Number: 769-39-1 EINECS: 212-209-6 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...