(1S)-(-)-Borneol 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12684-0B
A12684-18
A12684-30
CAAAA12684-0BEA
688.88
CAD
CAAAA12684-0B
CAAAA12684-18
CAAAA12684-30
(1S)-(-)-Borneol 97+%
(1S)-(-)-Borneol
Purity: 97+%
CAS No.: 464-45-9
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 65°(149°F)
Physical Form: Cryst solid
Melting Point: 208-211°
Boiling Point: 210°
Beilstein Reference: 3587558
Merck Reference: 14,1338
Optical Rotation: -36° (c=5 in ethanol)
UN No.: 1312
MDL No.: MFCD00003759
Formula:
C₁₀H₁₈O Boiling Pt: 210 °C Melting Pt: 208…211 °C Density: 1.011 g/cm³ (20 °C) Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003759 CAS Number: 464-45-9 EINECS: 207-353-1 UN: 1312 ADR: 4.1,III Merck Index: 14,1338 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...