α-Chloro-3-fluorotoulene 97%

Supplier: Thermo Scientific Chemicals
Danger

A12665-06 A12665-14 A12665-22
CAAAA12665-06EA 47.56 CAD
CAAAA12665-06 CAAAA12665-14 CAAAA12665-22
α-Chloro-3-fluorotoulene 97%
α-Chloro-3-fluorotoulene

Purity: 97% CAS No.: 456-42-8 Molecular Formula: C7H6ClF Formula Weight: 144.58 Flash Point: 58°(136°F) Refractive Index: 1.5120 Odor: Pungent Boiling Point: 176-177° Beilstein Reference: 742265 UN No.: 2920 MDL No.: MFCD00000904

Formula: C₇H₆ClF
Boiling Pt: 176…177 °C
Density: 1.213
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000904
CAS Number: 456-42-8
EINECS: 207-264-8
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR