1-Phenyl-2-tetrazoline-5-thione 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Mercapto-1-phenyl-1H-tetrazole, 1-Phenyl-5-mercaptotetrazole, 1-Phenyl-1H-tetrazole-5-thiol, 1-Phenyl-1H-tetrazole-5(2H)-thione

A12656-36 A12656-14 A12656-22
CAAAAA12656-36EA 1083.88 CAD
CAAAAA12656-36 CAAAA12656-14 CAAAA12656-22
1-Phenyl-2-tetrazoline-5-thione 99%
1-Phenyl-2-tetrazoline-5-thione

Purity: 99% CAS No.: 86-93-1 Molecular Formula: C7H6N4S Formula Weight: 178.22 Flash Point: 138°(280°F) Odor: Odorless Beilstein Reference: 139068 UN No.: 1325 MDL No.: MFCD00003129

Formula: C₇H₆N₄S
MW: 178.22 g/mol
Melting Pt: ∼ 145 °C (dec.)
Flash Pt: 138 °C (280 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003129
CAS Number: 86-93-1
EINECS: 201-710-5
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR