1-Butanesulfonic acid sodium salt 99%

Supplier: Thermo Scientific Chemicals
Warning

A12654-06 A12654-14 A12654-22
CAAAA12654-06EA 80.9 CAD
CAAAA12654-06 CAAAA12654-14 CAAAA12654-22
1-Butanesulfonic acid sodium salt 99%
1-Butansulphonic acid sodium salt

Purity: 99% CAS No.: 2386-54-1 Molecular Formula: CH3(CH2)3SO3Na Formula Weight: 160.17 Melting Point: 300° Beilstein Reference: 3716662

Formula: C₄H₉NaO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007540
CAS Number: 2386-54-1
EINECS: 219-201-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR