3-Bromo-4-methoxybenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12653-06
A12653-14
CAAAA12653-06EA
118.97
CAD
CAAAA12653-06
CAAAA12653-14
3-Bromo-4-methoxybenzoic acid 98+%
3-Bromo-4-methoxybenzoic acid
Purity: 98+%
CAS No.: 99-58-1
Molecular Formula: C8H7BrO3
Formula Weight: 231.05
Odor: Odorless
Melting Point: 217-222°
Beilstein Reference: 2087585
MDL No.: MFCD00020295
Formula:
C₈H₇BrO₃ Melting Pt: 217…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020295 CAS Number: 99-58-1 EINECS: 202-768-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...