2-Hexylthiophene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12652-09
A12652-18
CAAAA12652-09EA
161.16
CAD
CAAAA12652-09
CAAAA12652-18
2-Hexylthiophene 98%
2-Hexylthiophene
Purity: 98%
CAS No.: 18794-77-9
Molecular Formula: C10H16S
Formula Weight: 168.30
Flash Point: 100°(212°F)
Refractive Index: 1.4960
Boiling Point: 228-230°
Beilstein Reference: 110220
MDL No.: MFCD00022535
Formula:
C₁₀H₁₆S Boiling Pt: 228…230 °C Density: 0.935 Flash Pt: 100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00022535 CAS Number: 18794-77-9 EINECS: 242-579-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...